Tisk…

2-Amino-5-fluorophenylboronic acid

Dodavatel: Apollo Scientific

PC412217-100MG PC412217-250MG
APOSPC412217-100MGEA 5790 CZK
APOSPC412217-100MG APOSPC412217-250MG
2-Amino-5-fluorophenylboronic acid
2-Amino-5-fluorophenylboronic acid
Vzorec: C₆H₇BFNO₂
Storage Temperature: Ambient
MDL Number: MFCD03095362
CAS číslo: 1040400-87-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR