Tisk…

3-(2,2,2-Trifluoroethoxy)phenylboronic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-(2,2,2-Trifluoroethoxy)benzeneboronic acid, (3-(2,2,2-Trifluoroethoxy)phenyl)boronic acid

PC412086-1G PC412086-5G
APOSPC412086-1GEA 7050 CZK
APOSPC412086-1G APOSPC412086-5G
3-(2,2,2-Trifluoroethoxy)phenylboronic acid 97%
3-(2,2,2-Trifluoroethoxy)phenylboronic acid
Vzorec: C₈H₈BF₃O₃
MW: 219,95 g/mol
Bod tání: 126…129 °C
Storage Temperature: Ambient
MDL Number: MFCD07363846
CAS číslo: 850593-08-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR