N-Cyclopropyl-4-fluorobenzylamine 95+%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Cyclopropyl(4-fluorobenzyl)amine hydrochloride
PC411328-1G
PC411328-5G
APOSPC411328-1GEA
4960
CZK
APOSPC411328-1G
APOSPC411328-5G
N-Cyclopropyl-4-fluorobenzylamine 95+%
N-Cyclopropyl-4-fluorobenzylamine
Vzorec:
C₁₀H₁₂FN MW: 165,211 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07330007 CAS číslo: 625437-46-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...