4-Bromo-1-(difluoromethyl)-3-methyl-1H-pyrazole
Dodavatel: Apollo Scientific
|
Varování
|
PC411325-1G
PC411325-25G
PC411325-5G
APOSPC411325-1GEA
1780
CZK
APOSPC411325-1G
APOSPC411325-25G
APOSPC411325-5G
4-Bromo-1-(difluoromethyl)-3-methyl-1H-pyrazole
4-Bromo-1-(difluoromethyl)-3-methyl-1H-pyrazole
Vzorec:
C₅H₅BrF₂N₂ Storage Temperature: Ambient |
MDL Number:
MFCD14702889 CAS číslo: 1215295-92-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...