Tisk…

1-(Difluoromethyl)-4-iodo-5-methyl-1H-pyrazole

Dodavatel: Apollo Scientific
Varování

PC411324-10G PC411324-1G PC411324-5G
APOSPC411324-10GEA 16570 CZK
APOSPC411324-10G APOSPC411324-1G APOSPC411324-5G
1-(Difluoromethyl)-4-iodo-5-methyl-1H-pyrazole
1-(Difluoromethyl)-4-iodo-5-methyl-1H-pyrazole
Vzorec: C₅H₅F₂IN₂
Storage Temperature: Ambient
MDL Number: MFCD21364399
CAS číslo: 1354705-44-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR