Tisk…

2-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]ethanamine 95+%

Dodavatel: Apollo Scientific
Varování

PC411319-1G PC411319-5G
APOSPC411319-1GEA 1330 CZK
APOSPC411319-1G APOSPC411319-5G
2-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]ethanamine 95+%
2-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]ethanamine
Vzorec: C₇H₁₀F₃N₃
Storage Temperature: Ambient
MDL Number: MFCD04967724
CAS číslo: 925154-89-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR