Tisk…

2-(4-Fluorophenoxy)-3-nitropyridine 97%

Dodavatel: Apollo Scientific

PC4107F-1G PC4107F-5G
APOSPC4107F-1GEA 830 CZK
APOSPC4107F-1G APOSPC4107F-5G
2-(4-Fluorophenoxy)-3-nitropyridine 97%
2-(4-Fluorophenoxy)-3-nitropyridine
Vzorec: C₁₁H₇FN₂O₃
Bod tání: 98…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00067801
CAS číslo: 147143-58-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR