Tisk…

2,3-Difluorophenylboronic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: Dihydroxy(2,3-difluorophenyl)borane, 2,3-Difluorobenzeneboronic acid, (2,3-Difluorophenyl)metaboric acid

PC4104-100G PC4104-1G PC4104-25G
APOSPC4104-100GEA 9330 CZK
APOSPC4104-100G APOSPC4104-1G APOSPC4104-25G
2,3-Difluorophenylboronic acid 97%
2,3-Difluorophenylboronic acid
Vzorec: C₆H₅BF₂O₂
MW: 157,91 g/mol
Bod tání: 89 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01863170
CAS číslo: 121219-16-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR