Tisk…

1-Isothiocyanato-3-(trifluoromethoxy)benzene

Dodavatel: Apollo Scientific
Nebezpečí

PC410056-1G PC410056-25G PC410056-5G PC410056-100G
APOSPC410056-1GEA 1870 CZK
APOSPC410056-1G APOSPC410056-25G APOSPC410056-5G APOSPC410056-100G
1-Isothiocyanato-3-(trifluoromethoxy)benzene
1-Isothiocyanato-3-(trifluoromethoxy)benzene
Vzorec: C₈H₄F₃NOS
Storage Temperature: Ambient
MDL Number: MFCD07596437
CAS číslo: 471937-78-7
UN: 2922
ADR: 8+6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR