3-Fluoro-4-phenoxyaniline 98%
Dodavatel: Apollo Scientific
PC410050-10G
PC410050-5G
PC410050-1G
APOSPC410050-10GEA
11540
CZK
APOSPC410050-10G
APOSPC410050-5G
APOSPC410050-1G
3-Fluoro-4-phenoxyaniline 98%
3-Fluoro-4-phenoxyaniline
Vzorec:
C₁₂H₁₀FNO Storage Temperature: Ambient |
MDL Number:
MFCD08687878 CAS číslo: 39177-22-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...