1,6-Naphthyridin-5-yltrifluoromethanesulphonate 95+%
Dodavatel: Apollo Scientific
PC409511-1G
PC409511-10G
PC409511-5G
APOSPC409511-1GEA
13390
CZK
APOSPC409511-1G
APOSPC409511-10G
APOSPC409511-5G
1,6-Naphthyridin-5-yltrifluoromethanesulphonate 95+%
1,6-Naphthyridin-5-yltrifluoromethanesulphonate
Vzorec:
C₉H₅F₃N₂O₃S MW: 278,21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08567948 CAS číslo: 909649-09-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...