Tisk…

4-Fluoro-1-naphthaldehyde 95+%

Dodavatel: Apollo Scientific

PC409500-1G PC409500-25G PC409500-5G
APOSPC409500-1GEA 1660 CZK
APOSPC409500-1G APOSPC409500-25G APOSPC409500-5G
4-Fluoro-1-naphthaldehyde 95+%
4-Fluoro-1-naphthaldehyde
Vzorec: C₁₁H₇FO
MW: 174,1700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02261765
CAS číslo: 172033-73-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR