(4,4-Difluorocyclohexyl)methanamine hydrochloride 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC409007-1G
PC409007-5G
APOSPC409007-1GEA
3060
CZK
APOSPC409007-1G
APOSPC409007-5G
(4,4-Difluorocyclohexyl)methanamine hydrochloride 97%
(4,4-Difluorocyclohexyl)methanamine hydrochloride
Vzorec:
C₇H₁₄ClF₂N Storage Temperature: Ambient |
MDL Number:
MFCD13189562 CAS číslo: 809273-65-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...