Tisk…

1-Boc-3,3-difluoropyrrolidine

Dodavatel: Apollo Scientific

PC409004-1G PC409004-250MG
APOSPC409004-1GEA 2580 CZK
APOSPC409004-1G APOSPC409004-250MG
1-Boc-3,3-difluoropyrrolidine
1-Boc-3,3-difluoropyrrolidine
Vzorec: C₉H₁₅F₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD06797010
CAS číslo: 195447-25-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR