Tisk…

3-Amino-5-fluorobenzoic acid 97%

Dodavatel: Apollo Scientific

PC408286-25G
APOSPC408286-25GEA 6390 CZK
APOSPC408286-25G
3-Amino-5-fluorobenzoic acid 97%
3-Amino-5-fluorobenzoic acid
Vzorec: C₇H₆FNO₂
MW: 155,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07779526
CAS číslo: 786616-54-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR