Tisk…

(5-Fluoro-2-nitrophenyl)acetic acid

Dodavatel: Apollo Scientific

Synonyms: 5-Fluoro-2-nitrophenylacetic acid

PC408271-100G PC408271-25G
APOSPC408271-100GEA 11080 CZK
APOSPC408271-100G APOSPC408271-25G
(5-Fluoro-2-nitrophenyl)acetic acid
(5-Fluoro-2-nitrophenyl)acetic acid
Vzorec: C₈H₆FNO₄
MW: 199,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07787601
CAS číslo: 29640-98-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR