2,3,4,5-Tetrafluorobenzaldehyde
Dodavatel: Apollo Scientific
|
Varování
|
PC408137-5G
PC408137-1G
APOSPC408137-5GEA
9160
CZK
APOSPC408137-5G
APOSPC408137-1G
2,3,4,5-Tetrafluorobenzaldehyde
2,3,4,5-Tetrafluorobenzaldehyde
Vzorec:
C₇H₂F₄O MW: 178,09 g/mol Bod varu: 82 °C/62mmHg Hustota: 1,51 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD02683041 CAS číslo: 16583-06-5 UN: 1989 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...