Tisk…

6-Fluoroisatin

Dodavatel: Apollo Scientific

Synonyms: 6-Fluoro-1H-indole-2,3-dione, 6-Fluoroindoline-2,3-dione

PC408008-10G PC408008-1G PC408008-5G
APOSPC408008-10GEA 22780 CZK
APOSPC408008-10G APOSPC408008-1G APOSPC408008-5G
6-Fluoroisatin
6-Fluoroisatin
Vzorec: C₈H₄FNO₂
MW: 165,12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03618556
CAS číslo: 324-03-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR