2-Fluoro-4-(pentafluorothio)aniline 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC405669-1G
PC405669-100MG
APOSPC405669-1GEA
16370
CZK
APOSPC405669-1G
APOSPC405669-100MG
2-Fluoro-4-(pentafluorothio)aniline 97%
2-Fluoro-4-(pentafluorothio)aniline
Vzorec:
C₆H₅F₆NS MW: 237,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16652511 CAS číslo: 1240257-25-3 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...