Tisk…

3-Fluoro-5-(pentafluorothio)benzaldehyde 97%

Dodavatel: Apollo Scientific
Varování

PC405666-1G PC405666-5G PC405666-10G
APOSPC405666-1GEA 9920 CZK
APOSPC405666-1G APOSPC405666-5G APOSPC405666-10G
3-Fluoro-5-(pentafluorothio)benzaldehyde 97%
3-Fluoro-5-(pentafluorothio)benzaldehyde
Vzorec: C₇H₄F₆OS
MW: 250,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16652417
CAS číslo: 1240257-92-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR