Tisk…

2-Fluoro-5-(pentafluorothio)aniline 98%

Dodavatel: Apollo Scientific
Varování

PC405595-1G PC405595-5G
APOSPC405595-1GEA 12010 CZK
APOSPC405595-1G APOSPC405595-5G
2-Fluoro-5-(pentafluorothio)aniline 98%
2-Fluoro-5-(pentafluorothio)aniline
Vzorec: C₆H₅F₆NS
MW: 237,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16652446
CAS číslo: 1240257-94-6
UN: 2810
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR