3-Bromo-5-(pentafluorothio)aniline 97%
Dodavatel: Apollo Scientific
PC405594-1G
PC405594-5G
PC405594-10G
APOSPC405594-1GEA
12010
CZK
APOSPC405594-1G
APOSPC405594-5G
APOSPC405594-10G
3-Bromo-5-(pentafluorothio)aniline 97%
3-Bromo-5-(pentafluorothio)aniline
Vzorec:
C₆H₅BrF₅NS MW: 298,07 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16652416 CAS číslo: 1240257-95-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...