5-Fluoroorotic acid monohydrate (5-FOA) 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-FOA
PC4054-25G
PC4054-5G
APOSPC4054-25GEA
12010
CZK
APOSPC4054-25G
APOSPC4054-5G
5-Fluoroorotic acid monohydrate (5-FOA) 98%
5-Fluoroorotic acid monohydrate (5-FOA)
Useful in the selection of orotidine-5'-phosphatedecarboxylase mutants in Saccharomycescerevisiae: Genetics, 1984, 107, 179.
Vzorec:
C₅H₃FN₂O₄·1H₂O MW: 174.0 g/mol (anhy) Bod tání: 255 °C (rozkl.) Storage Temperature: Freezer |
MDL Number:
MFCD00150658 CAS číslo: 220141-70-8 EINECS: 211-876-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...