Tisk…

trans-tert-Butyl 3-fluoro-4-hydroxypyrrolidine-1-carboxylate

Dodavatel: Apollo Scientific
Varování

Synonyms: tert-Butyl trans-3-fluoro-4-hydroxypyrrolidine-1-carboxylate, trans-3-Fluoro-4-hydroxypyrrolidine, N-BOC protected

PC405008-1G PC405008-5G
APOSPC405008-1GEA 6020 CZK
APOSPC405008-1G APOSPC405008-5G
trans-tert-Butyl 3-fluoro-4-hydroxypyrrolidine-1-carboxylate
trans-tert-Butyl 3-fluoro-4-hydroxypyrrolidine-1-carboxylate
Vzorec: C₉H₁₆FNO₃
MW: 205,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12032106
CAS číslo: 869481-93-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR