Tisk…

4-Bromo-6-fluoro-2-methylquinoline

Dodavatel: Apollo Scientific

PC404614-1G PC404614-5G
APOSPC404614-1GEA 6910 CZK
APOSPC404614-1G APOSPC404614-5G
4-Bromo-6-fluoro-2-methylquinoline
4-Bromo-6-fluoro-2-methylquinoline
Vzorec: C₁₀H₇BrFN
Storage Temperature: Ambient
MDL Number: MFCD11505132
CAS číslo: 1070879-47-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR