Tisk…

Mefloquine hydrochloride

Dodavatel: Apollo Scientific
Varování

Synonyms: (S)-[2,8-Bis(trifluoromethyl)quinolin-4-yl][(2R)-piperidin-2-yl]methanol hydrochloride, rel-(S)-(2,8-Bis(trifluoromethyl)quinolin-4-yl)((R)-piperidin-2-yl)methanol hydrochloride, Mefloquine HCI

PC404546-1G PC404546-5G
APOSPC404546-1GEA 3190 CZK
APOSPC404546-1G APOSPC404546-5G
Mefloquine hydrochloride
Mefloquine hydrochloride
Vzorec: C₁₇H₁₆F₆N₂O
MW: 414,78 g/mol
Bod tání: 259…264 °C
Storage Temperature: Ambient
MDL Number: MFCD00797519
CAS číslo: 51773-92-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR