Tisk…

Methyl-2-[2-(trifluoromethyl)quinolin-4-ylthio]acetate

Dodavatel: Apollo Scientific

PC4043-1G PC4043-5G
APOSPC4043-1GEA 1870 CZK
APOSPC4043-1G APOSPC4043-5G
Methyl-2-[2-(trifluoromethyl)quinolin-4-ylthio]acetate
Methyl-2-[2-(trifluoromethyl)quinolin-4-ylthio]acetate
Vzorec: C₁₃H₁₀F₃NO₂S
MW: 301,29 g/mol
Bod tání: 96…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00219657
CAS číslo: 175203-42-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR