Tisk…

(2-Fluoropyridin-3-yl)methanamine 95+%

Dodavatel: Apollo Scientific

PC403116-250MG PC403116-1G
APOSPC403116-250MGEA 3020 CZK
APOSPC403116-250MG APOSPC403116-1G
(2-Fluoropyridin-3-yl)methanamine 95+%
(2-Fluoropyridin-3-yl)methanamine
Vzorec: C₆H₇FN₂
MW: 126,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06213866
CAS číslo: 205744-16-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR