Tisk…

3-(4-Fluorophenyl)piperidine hydrochloride

Dodavatel: Apollo Scientific

PC402142-1G PC402142-250MG
APOSPC402142-1GEA 15940 CZK
APOSPC402142-1G APOSPC402142-250MG
3-(4-Fluorophenyl)piperidine hydrochloride
3-(4-Fluorophenyl)piperidine hydrochloride
Vzorec: C₁₁H₁₅ClFN
Storage Temperature: Refrigerator
MDL Number: MFCD02178931
CAS číslo: 1106940-94-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR