Linifanib
Dodavatel: Apollo Scientific
PC402063-100MG
PC402063-1G
PC402063-250MG
APOSPC402063-100MGEA
5430
CZK
APOSPC402063-100MG
APOSPC402063-1G
APOSPC402063-250MG
Linifanib
Linifanib
Vzorec:
C₂₁H₁₈FN₅O Storage Temperature: Ambient |
MDL Number:
MFCD11840918 CAS číslo: 796967-16-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...