Tisk…

1-(Trifluoromethyl)cyclopropanecarboxylic acid 99%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1-(Trifluoromethyl)cyclopropane-1-carboxylic acid

PC4014-1G PC4014-5G PC4014-25G
APOSPC4014-1GEA 744 CZK
APOSPC4014-1G APOSPC4014-5G APOSPC4014-25G
1-(Trifluoromethyl)cyclopropanecarboxylic acid 99%
1-(Trifluoromethyl)cyclopropanecarboxylic acid
Vzorec: C₅H₅F₃O₂
MW: 154,09 g/mol
Bod varu: 182…184 °C
Bod tání: 86…89 °C
Storage Temperature: Ambient
MDL Number: MFCD03093070
CAS číslo: 277756-46-4
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR