Tisk…

2-Phenyl-5-trifluoromethyl-2H-pyrazol-3-ol

Dodavatel: Apollo Scientific

PC401047-1G PC401047-5G
APOSPC401047-1GEA 695 CZK
APOSPC401047-1G APOSPC401047-5G
2-Phenyl-5-trifluoromethyl-2H-pyrazol-3-ol
2-Phenyl-5-trifluoromethyl-2H-pyrazol-3-ol
Vzorec: C₁₀H₇F₃N₂O
Storage Temperature: Ambient
MDL Number: MFCD03714773
CAS číslo: 96145-98-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR