Tisk…

2-Bromo-5-fluoro-3-nitropyridine

Dodavatel: Apollo Scientific
Varování

PC400737-25G PC400737-5G
APOSPC400737-25GEA 9100 CZK
APOSPC400737-25G APOSPC400737-5G
2-Bromo-5-fluoro-3-nitropyridine
2-Bromo-5-fluoro-3-nitropyridine
Vzorec: C₅H₂BrFN₂O₂
MW: 220.99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05662387
CAS číslo: 652160-72-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR