Methyl-3-fluoro-2-nitrobenzoate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Methyl 3-fluoro-2-nitrobenzoate
PC400724-1G
PC400724-25G
PC400724-5G
APOSPC400724-1GEA
1450
CZK
APOSPC400724-1G
APOSPC400724-25G
APOSPC400724-5G
Methyl-3-fluoro-2-nitrobenzoate
Methyl-3-fluoro-2-nitrobenzoate
Vzorec:
C₈H₆FNO₄ MW: 199,1360 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11520393 CAS číslo: 1214353-57-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...