Tisk…

3,3-Difluorocyclobutanecarboxylic acid 98%

Dodavatel: Apollo Scientific
Varování

PC400716-10G PC400716-1G PC400716-5G
APOSPC400716-10GEA 4140 CZK
APOSPC400716-10G APOSPC400716-1G APOSPC400716-5G
3,3-Difluorocyclobutanecarboxylic acid 98%
3,3-Difluorocyclobutanecarboxylic acid
Vzorec: C₅H₆F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD08705857
CAS číslo: 107496-54-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR