5-Fluoropyrimidin-4-amine
Dodavatel: Apollo Scientific
|
Varování
|
PC400708-1G
PC400708-250MG
PC400708-5G
APOSPC400708-1GEA
5430
CZK
APOSPC400708-1G
APOSPC400708-250MG
APOSPC400708-5G
5-Fluoropyrimidin-4-amine
5-Fluoropyrimidin-4-amine
Vzorec:
C4H4FN3 MW: 113,0931 g/mol Bod varu: 222,7 °C/760mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00223685 CAS číslo: 811450-26-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...