Tisk…

3-Fluoro-5-hydroxybenzonitrile 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC400705-25G
APOSPC400705-25GEA 3380 CZK
APOSPC400705-25G
3-Fluoro-5-hydroxybenzonitrile 97%
3-Fluoro-5-hydroxybenzonitrile
Vzorec: C₇H₄FNO
MW: 137,1112 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09972133
CAS číslo: 473923-95-4
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR