Tisk…

4-(4-Trifluoromethylpyridin-2-yl)benzoic acid

Dodavatel: Apollo Scientific

PC400664-1G PC400664-250MG PC400664-500MG
APOSPC400664-1GEA 8400 CZK
APOSPC400664-1G APOSPC400664-250MG APOSPC400664-500MG
4-(4-Trifluoromethylpyridin-2-yl)benzoic acid
4-(4-Trifluoromethylpyridin-2-yl)benzoic acid
Vzorec: C₁₃H₈F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD18384864
CAS číslo: 1299607-71-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR