(1R,2R)-2-Fluorocyclopropanecarboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
PC400536-1G
PC400536-250MG
APOSPC400536-1GEA
2070
CZK
APOSPC400536-1G
APOSPC400536-250MG
(1R,2R)-2-Fluorocyclopropanecarboxylic acid
(1R,2R)-2-Fluorocyclopropanecarboxylic acid
Vzorec:
C₄H₅FO₂ MW: 104.08 g/mol Bod tání: 55…56 °C Storage Temperature: Ambient |
MDL Number:
MFCD19238080 CAS číslo: 127199-13-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...