Tisk…

tert-Butyl-3,5-difluoropyridin-4-ylcarbamate

Dodavatel: Apollo Scientific

Synonyms: 4-Amino-3,5-difluoropyridine, 4-BOC protected

PC400500-1G PC400500-250MG
APOSPC400500-1GEA 20500 CZK
APOSPC400500-1G APOSPC400500-250MG
tert-Butyl-3,5-difluoropyridin-4-ylcarbamate
tert-Butyl-3,5-difluoropyridin-4-ylcarbamate
Vzorec: C₁₀H₁₂F₂N₂O₂
MW: 230.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22199252
CAS číslo: 1364663-23-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR