4-Fluoro-3-nitrophenyl isocyanate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-Fluoro-4-isocyanato-2-nitrobenzene
PC3985-5G
PC3985-1G
APOSPC3985-5GEA
16340
CZK
APOSPC3985-5G
APOSPC3985-1G
4-Fluoro-3-nitrophenyl isocyanate
4-Fluoro-3-nitrophenyl isocyanate
When melted, may contain small amount of sediment.
Vzorec:
C₇H₃FN₂O₃ Bod tání: 27…28 °C Hustota: 1,455 g/cm³ (25 °C) Bod vzplanutí: 113 °C Storage Temperature: Ambient |
CAS číslo:
65303-82-4 UN: 2206 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...