2-Nitro-5-(trifluoromethyl)aniline
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
3-Amino-4-nitrobenzotrifluoride
PC3972-1G
PC3972-5G
PC3972-10G
APOSPC3972-1GEA
2480
CZK
APOSPC3972-1G
APOSPC3972-5G
APOSPC3972-10G
2-Nitro-5-(trifluoromethyl)aniline
2-Nitro-5-(trifluoromethyl)aniline
Vzorec:
C₇H₅F₃N₂O₂ MW: 206,13 g/mol Bod tání: 98…102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042447 CAS číslo: 402-14-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...