3-Bromo-5-(trifluoromethoxy)benzyl bromide 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC3957-10G
PC3957-5G
PC3957-1G
APOSPC3957-10GEA
11180
CZK
APOSPC3957-10G
APOSPC3957-5G
APOSPC3957-1G
3-Bromo-5-(trifluoromethoxy)benzyl bromide 97%
3-Bromo-5-(trifluoromethoxy)benzyl bromide
Vzorec:
C₈H₅Br₂F₃O MW: 333.93 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11035864 CAS číslo: 1031929-10-8 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...