Tisk…

5-Carboxy-2-chloro-4-fluorophenylboronic acid

Dodavatel: Apollo Scientific

PC3955-1G PC3955-250MG
APOSPC3955-1GEA 14190 CZK
APOSPC3955-1G APOSPC3955-250MG
5-Carboxy-2-chloro-4-fluorophenylboronic acid
5-Carboxy-2-chloro-4-fluorophenylboronic acid
Vzorec: C₇H₅BClFO₄
Bod tání: 221…223 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09878325
CAS číslo: 957066-06-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR