Tisk…

4-Fluoro-N-isopropylbenzamide

Dodavatel: Apollo Scientific

PC3954-5G PC3954-1G PC3954-25G
APOSPC3954-5GEA 3310 CZK
APOSPC3954-5G APOSPC3954-1G APOSPC3954-25G
4-Fluoro-N-isopropylbenzamide
4-Fluoro-N-isopropylbenzamide
Vzorec: C₁₀H₁₂FNO
Bod tání: 142…144 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01214150
CAS číslo: 70001-45-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR