Tisk…

Ethyl-3-(2,6-dichloro-5-fluoro-3-pyridyl)-3-oxopropionate 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: (2,6-Dichloro-5-fluoronicotinoyl)acetic Acid Ethyl Ester, Ethyl (2,6-Dichloro-5-fluoronicotinoyl)acetate, Ethyl 3-(2,6-Dichloro-5-fluoro-3-pyridyl)-3-oxopropionate, 3-(2,6-Dichloro-5-fluoro-3-pyridyl)-3-oxopropionic Acid Ethyl Ester

PC3939-25G PC3939-5G PC3939-100G
APOSPC3939-25GEA 827 CZK
APOSPC3939-25G APOSPC3939-5G APOSPC3939-100G
Ethyl-3-(2,6-dichloro-5-fluoro-3-pyridyl)-3-oxopropionate 97%
Ethyl-3-(2,6-dichloro-5-fluoro-3-pyridyl)-3-oxopropionate
Vzorec: C₁₀H₈Cl₂FNO₃
MW: 280,08 g/mol
Bod tání: 66…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00799535
CAS číslo: 96568-04-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR