3-Bromo-5-fluorobenzoic acid 99%
Dodavatel: Apollo Scientific
|
Varování
|
PC3919-25G
PC3919-5G
PC3919-100G
APOSPC3919-25GEA
744
CZK
APOSPC3919-25G
APOSPC3919-5G
APOSPC3919-100G
3-Bromo-5-fluorobenzoic acid 99%
3-Bromo-5-fluorobenzoic acid
Vzorec:
C₇H₄BrFO₂ MW: 219,01 g/mol Bod tání: 141…145 °C Storage Temperature: Ambient |
MDL Number:
MFCD01569540 CAS číslo: 176548-70-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...