1-Fluoro-2-nitrobenzene 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
o-Fluoronitrobenzene, 2-Fluoronitrobenzene
PC3890-1KG
PC3890-250G
PC3890-25G
APOSPC3890-1KGEA
3610
CZK
APOSPC3890-1KG
APOSPC3890-250G
APOSPC3890-25G
1-Fluoro-2-nitrobenzene 98%
1-Fluoro-2-nitrobenzene
The activated fluorine reacts with arylamines to form 2-nitrodiarylamines see: Synthesis, 215 (1980)
Vzorec:
C₆H₄FNO₂ MW: 141,1 g/mol Bod varu: 116 °C/22mmHg Bod tání: –6 °C Hustota: 1,338 g/cm³ (25 °C) Bod vzplanutí: 94 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007048 CAS číslo: 1493-27-2 EINECS: 216-088-0 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...