2,2,2-Trifluoroethyl methanesulphonate 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC3889-5G
PC3889-1G
PC3889-25G
APOSPC3889-5GEA
2070
CZK
APOSPC3889-5G
APOSPC3889-1G
APOSPC3889-25G
2,2,2-Trifluoroethyl methanesulphonate 98%
2,2,2-Trifluoroethyl methanesulphonate
Vzorec:
C₃H₅F₃O₃S Hustota: 1,513 g/cm³ (25 °C) Bod vzplanutí: 104 °C Storage Temperature: Ambient |
MDL Number:
MFCD00025058 CAS číslo: 25236-64-0 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...