Tisk…

2,2,3-Trichloro-1,1,1,4,4,4-hexafluorobutane 97%

Dodavatel: Apollo Scientific

Synonyms: 1,1,1,4,4,4-Hexafluoro-2,2,3-trichlorobutane

PC3887-5G PC3887-1G PC3887-25G
APOSPC3887-5GEA 6850 CZK
APOSPC3887-5G APOSPC3887-1G APOSPC3887-25G
2,2,3-Trichloro-1,1,1,4,4,4-hexafluorobutane 97%
2,2,3-Trichloro-1,1,1,4,4,4-hexafluorobutane
Vzorec: C₄HCl₃F₆
MW: 269,4 g/mol
Bod varu: 105 °C
Hustota: 1,697 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD02093295
CAS číslo: 378-84-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR